Cholinchlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2-Hydroxyethyl)trimethylammoniumchlorid
A15828.22
A15828.0E
A15828.36
A15828.22EA
30.1
EUR
A15828.22
A15828.0E
A15828.36
Cholinchlorid ≥98%
Cholinchlorid
Purity: 98+%
CAS No.: 67-48-1
Molecular Formula: HOCH2CH2N(CH3)3Cl
Formula Weight: 139.63
Physical Form: Solid
Sensitivity: Hygroscopic
Beilstein Reference: 3563126
Merck Reference: 14,2206
MDL No.: MFCD00011721
An acyl group acceptor
Formel:
HOH₂CCH₂N(CH₃)₃Cl Molecular Weight: 139.62 g/mol Schmelzpunkt: ∼ 305 °C (Zers.) Dichte: ∼1.205 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011721 CAS-Nummer: 67-48-1 EINECS: 200-655-4 Merck Index: 13,02226 |