Acetylcholiniodid 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2-Acetoxyethyl)trimethylammoniumiodid, Acetylcholiniumiodid
400180100
400180500
ACRO400180100EA
71.9
EUR
ACRO400180100
ACRO400180500
Acetylcholiniodid 99%
Acetylcholiniodid
Formel:
H₃CC(O)O(CH₂)₂N(CH₃)₃I Molecular Weight: 273,11 g/mol Schmelzpunkt: 162…164 °C Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00011815 CAS-Nummer: 2260-50-6 EINECS: 218-862-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 156 to 164 °C |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...