Butyrylthiocholiniodid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Butyrylthiocholiniumiodid, (Propylcarbonylthioethyl)trimethylammoniumiodid, (2-(Butyrylthio)ethyl)trimethylammoniumiodid, (2-Butyrylthioethyl)trimethylammoniumiodid

108150250 108151000
ACRO108150250EA 178 EUR
ACRO108150250 ACRO108151000
Butyrylthiocholiniodid 98%
Butyrylthiocholiniodid
Formel: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
Molecular Weight: 317,23 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011845
CAS-Nummer: 1866-16-6
EINECS: 217-475-7

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration Argentometric ≥97.5 %
UV at 250 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR