Acetylthiocholiniodid
Lieferant: Serva
|
Gefahr
|
Synonyme:
(2-Acetylthioethyl)trimethylammoniumiodid, Acetylthiocholiniumiodid
10570.01
10570.02
10570.03
SERA10570.01EA
42.3
EUR
SERA10570.01
SERA10570.02
SERA10570.03
Acetylthiocholiniodid
Acetylthiocholiniodid
Formel:
H₃CC(S)O(CH₂)₂N(CH₃)₃I Molecular Weight: 289,18 g/mol Schmelzpunkt: 204 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00011819 CAS-Nummer: 1866-15-5 EINECS: 217-474-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...