Nitroniumtetrafluoroborat 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Nitryl tetrafluoroborate

B20167.06 B20167.14 B20167.22
B20167.06EA 63.7 EUR
B20167.06 B20167.14 B20167.22
Nitroniumtetrafluoroborat 96%
Nitroniumtetrafluoroborat

Purity: 96% CAS No.: 13826-86-3 Molecular Formula: BF4NO2 Formula Weight: 132.81 Storage Temperature: 2°C to 8°C Melting Point: ca 240° dec. UN No.: 3260 MDL No.: MFCD00011432

Formel: BF₄NO₂
Molecular Weight: 132,81 g/mol
Schmelzpunkt: ∼ 240 °C (Zersetzung)
Dichte: 1,26 g/cm³ (20 °C)
MDL: MFCD00011432
CAS-Nummer: 13826-86-3
EINECS: 237-533-5
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR