Chlordiphenylphosphin 95% tech.
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
DPPC
109130250
109131000
109135000
ACRO109130250EA
43.6
EUR
ACRO109130250
ACRO109131000
ACRO109135000
Chlordiphenylphosphin 95% tech.
Chlordiphenylphosphin
Formel:
C₁₂H₁₀ClP Molecular Weight: 220,64 g/mol Siedepunkt: 320 °C (760 mmHg) Schmelzpunkt: 14…16 °C Dichte: 1,229 g/cm³ Flammpunkt: 138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000529 CAS-Nummer: 1079-66-9 EINECS: 214-093-2 UN: 3265 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Yellow Liquid (May develop some turbidity or precipitate) |
Infrared spectrum | Conforms |
Refractive index | 1.6340 to 1.6370 (20°C, 589 nm) |
GC | ≥94 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Carbonyl compounds | ≤0.002 % (Acetone) |
Titrable acid or base | ≤0.0001 meq/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...