Diphenylphosphin 95%

Lieferant: Thermo Scientific
Gefahr

320970050 320970250
ACRO320970050EA 75 EUR
ACRO320970050 ACRO320970250
Diphenylphosphin 95%
Diphenylphosphin
Formel: C₁₂H₁₁P
Molecular Weight: 186,19 g/mol
Flammpunkt: 110 °C
MDL: MFCD00003040
CAS-Nummer: 829-85-6
EINECS: 212-591-4
UN: 2845
ADR: 4.2,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6250 to 1.6300 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR