Ammoniumtetrafluoroborat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate
11488.36
11488.A3
11488.36EA
58
EUR
11488.36
11488.A3
Ammoniumtetrafluoroborat ≥97%
Ammonium tetrafluoroborate
Purity: 97%
CAS No.: 13826-83-0
Molecular Formula: NH4BF4
Formula Weight: 104.84
Physical Form: Powder
Density: 1.871
Appearance: White
Odour: Ammonia-like
Melting Point: 220° subl.
UN No.: 3260
MDL No.: MFCD06245785
Formel:
H₄BF₄N Schmelzpunkt: 220 °C (Subl.) Dichte: 1,871 Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245785 CAS-Nummer: 13826-83-0 EINECS: 237-531-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...