Ammoniumbromid ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Achtung
12981.06 12981.14 12981.22
12981.06EA 39.3 EUR
12981.06 12981.14 12981.22
Ammoniumbromid ≥99,999% (Metall-Basis), Puratronic®
Ammonium bromide
Purity: 99.999% CAS No.: 12124-97-9 Molecular Formula: NH4Br Formula Weight: 97.97 Physical Form: Crystalline Density: 2.429 Appearance: White Melting Point: 452° subl. Merck Reference: 14,505 MDL No.: MFCD00011419
Formel: H₄BrN
Molecular Weight: 97,94 g/mol
Schmelzpunkt: 452 °C (Subl.)
Dichte: 2,429
Lagertemperatur: Raumtemperatur
MDL: MFCD00011419
CAS-Nummer: 12124-97-9
EINECS: 235-183-8
Merck Index: 14,505