Ammoniumbromid, reinst

Lieferant: Bernd Kraft

15185.3600 15185.5600 15185.1600 15185.2600
KRAF15185.3600EA 78.3 EUR
KRAF15185.3600 KRAF15185.5600 KRAF15185.1600 KRAF15185.2600
Ammoniumbromid, reinst
Ammonium bromide
Formel: H₄BrN
Molecular Weight: 97,94 g/mol
Schmelzpunkt: 542 °C
Dichte: 2,42 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011419
CAS-Nummer: 12124-97-9
EINECS: 235-183-8
Merck Index: 13,00505

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR