Ammoniumchlorid ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Salmiak
10632.18
10632.30
10632.A1
10632.18EA
89
EUR
10632.18
10632.30
10632.A1
Ammoniumchlorid ≥99,999% (Metall-Basis), Puratronic®
Ammoniumchlorid
Purity: 99.999%
CAS No.: 12125-02-9
Molecular Formula: NH4Cl
Formula Weight: 53.49
Physical Form: Powder
Density: ≈1.53
Refractive Index: 1.642
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 340° subl.
Merck Reference: 14,509
MDL No.: MFCD00011420
Formel:
NH₄Cl Molecular Weight: 53,49 g/mol Schmelzpunkt: 340 °C (Subl.) Dichte: 1,53 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011420 CAS-Nummer: 12125-02-9 EINECS: 235-186-4 Merck Index: 14,509 |