Ammoniumcarbonat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Achtung
Synonyme: Ammoniumcarbonat, Hirschhornsalz
12980.09 12980.18 12980.30
12980.09EA 29.7 EUR
12980.09 12980.18 12980.30
Ammoniumcarbonat ≥99,999% (Metall-Basis), Puratronic®
Ammoniumcarbonat (enthält Ammoniumcarbamat)
Purity: 99.999% CAS No.: 506-87-6 Molecular Formula: CH8N2O3 Formula Weight: 96.09 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Ammonia-like Melting Point: 60° subl. Beilstein Reference: 3627235 Merck Reference: 14,508 MDL No.: MFCD00010890
Formel: (NH₄)₂CO₃
Molecular Weight: 96,09 g/mol
Schmelzpunkt: 60 °C (Subl.)
Dichte: 1,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010890
CAS-Nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 14,508