Ammoniumfluorid 98+%, reinst
Lieferant: Thermo Scientific
|
Gefahr
|
ACRO201845000EA
96.4
EUR
ACRO201845000
ACRO201841000
ACRO201840025
Ammoniumfluorid 98+%, reinst
Ammonium fluoride
Formel:
NH₄F Molecular Weight: 37,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011423 CAS-Nummer: 12125-01-8 EINECS: 235-185-9 UN: 2505 ADR: 6.1,III Merck Index: 13,00524 |
Spezifikation Testergebnisse
Appearance | White Adhering crystalline powder |
Residue after ignition | ≤0.01 % (SO4) |
Titration excess NaOH | ≥98.0 % |
Hexafluorosilicate (SiF6) | ≤0.1 % |
Heavy metals (as Pb) | ≤5 ppm |
(NH4)HF2 | ≤1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...