Ammoniumperoxodisulfat (APS) ≥98%, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Peroxidisulphuric acid diammonium salt, Ammonium persulphate
31117-1KG
31117-50KG
31117-5KG
31117-1KGEA
68.28
EUR
31117-1KG
HONC31117-50KG
31117-5KG
Ammoniumperoxodisulfat (APS) ≥98%, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™
Ammoniumperoxodisulfat (APS)
Formel:
(NH₄)₂S₂O₈ Molecular Weight: 228.2 g/mol Schmelzpunkt: 120 Dichte: 1.98 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003390 CAS-Nummer: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1 III Merck Index: 13,00545 |
|
Spezifikation Testergebnisse
Assay | Min. 98 % |
Water insoluble matter | Max. 0.005 % |
Residue on ignition (SO₄) | Max. 0.05 % |
Titrable Acid | Max. 0.04 meq/g |
Iron (Fe) | Max. 0.001 % |
Manganese (Mn) | Max. 0.00005 % |
Heavy metals (as Pb) | Max. 0.001 % |
Chloride, Chlorate (as Cl) | Max. 0.001 % |
Chloride (Cl) | Max. 0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...