Ammoniumhexafluorosilicat
Lieferant: CHEMPUR SILANE
|
Gefahr
|
Synonyme:
Ammoniumfluorosilicat
SILS00875.250
SILS00875.1K
SILNSILS00875.250EA
0
EUR
SILNSILS00875.250
SILNSILS00875.1K
Ammoniumhexafluorosilicat
Ammoniumhexafluorosilicat
Formel:
H₈F₆N₂Si Molecular Weight: 178,15 g/mol Dichte: 2,01 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010887 CAS-Nummer: 16919-19-0 EINECS: 240-968-3 UN: 2854 ADR: 6.1,III Merck Index: 13,00529 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...