Ammoniumsulfat
Lieferant: Serva
Synonyme:
Diammoniumsulfat, di-Ammoniumsulfat
13385.01
13385.02
SERA13385.01EA
0
EUR
SERA13385.01
SERA13385.02
Ammoniumsulfat
Ammoniumsulfat
Formel:
(NH₄)₂SO₄ Molecular Weight: 132.14 g/mol Schmelzpunkt: 336…339 °C Dichte: 1.78 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...