(Diethylamino)schwefeltrifluorid
Lieferant: FLUOROCHEM
Synonyme:
DAST
002323-25G
002323-100G
FLUO002323-25GEA
145
EUR
FLUO002323-25G
FLUO002323-100G
(Diethylamino)schwefeltrifluorid
(Diethylamino)schwefeltrifluorid
Formel:
C₄H₁₀F₃NS Molecular Weight: 161,19 g/mol Dichte: 1,316 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000363 CAS-Nummer: 38078-09-0 EINECS: 253-771-2 UN: 2920 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...