Diallylsulfid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Allyl sulphide

L10971.14 L10971.22
L10971.14EA 35.4 EUR
L10971.14 L10971.22
Diallylsulfid ≥98%
Diallylsulfid

Purity: 98% CAS No.: 592-88-1 Molecular Formula: C6H10S Formula Weight: 114.21 Flash Point: 23°(73°F) Refractive Index: 1.4895 Odour: Unpleasant Melting Point: -83° Boiling Point: 137-138° Beilstein Reference: 1736016 Merck Reference: 14,297 UN No.: 1993 MDL No.: MFCD00008658

Formel: C₆H₁₀S
Molecular Weight: 114,21 g/mol
Siedepunkt: 137…138 °C
Schmelzpunkt: –83 °C
Dichte: 0,889
Flammpunkt: 23 °C (73 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008658
CAS-Nummer: 592-88-1
EINECS: 209-775-1
UN: 1993
ADR: 3,III
Merck Index: 14,297

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR