Disulfiram 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tetraethylthiuramdisulfid, Bis(diethyl-thiocarbamoyl)disulfid
138120250
138125000
138120025
ACRO138120250EA
19.2
EUR
ACRO138120250
ACRO138125000
ACRO138120025
Disulfiram 97%
Disulfiram
Formel:
C₁₀H₂₀N₂S₄ Molecular Weight: 296.55 g/mol Siedepunkt: 117 °C (23hPa) Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009048 CAS-Nummer: 97-77-8 EINECS: 202-607-8 UN: 3077 ADR: 9 III Merck Index: 13,03399 |
Spezifikation Testergebnisse
Appearance | White to cream to light yellow Crystalline powder or crystals or granules |
Infrared spectrum | Conforms |
Melting point | 67 to 74 °C |
Water | ≤1.0 % (KF) |
HPLC | ≥96.0 % |
Ash | ≤0.3 % (900°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...