Disulfiram ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tetraethylthiuramdisulfid, Bis(diethyl-thiocarbamoyl)disulfid

B20721.0B B20721.0I B20721.22 B20721.30
B20721.0BEA 110 EUR
B20721.0B B20721.0I B20721.22 B20721.30
Disulfiram ≥97%
Disulfiram

Purity: 97% CAS No.: 97-77-8 Molecular Formula: C10H20N2S4 Formula Weight: 296.54 Melting Point: 69-72° Boiling Point: 117°/17mm Merck Reference: 14,3364 UN No.: 3077 MDL No.: MFCD00009048

Formel: C₁₀H₂₀N₂S₄
Siedepunkt: 117 °C (17 mmHg)
Schmelzpunkt: 69…72 °C
Dichte: 1,300
Lagertemperatur: Raumtemperatur
MDL: MFCD00009048
CAS-Nummer: 97-77-8
EINECS: 202-607-8
UN: 3077
ADR: 9,III
Merck Index: 14,3364

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR