Thiram 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Bis(dimethyl-thiocarbamoyl)disulfid

138480010 138480050 138485000
ACRO138480010EA 97.9 EUR
ACRO138480010 ACRO138480050 ACRO138485000
Thiram 97%
Thiram
Formel: C₆H₁₂N₂S₄
Molecular Weight: 240,44 g/mol
Siedepunkt: 129 °C (20 mmHg)
Schmelzpunkt: >145 °C
Dichte: 1,426 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00008325
CAS-Nummer: 137-26-8
EINECS: 205-286-2
UN: 2811
ADR: 6.1,II
Merck Index: 12,09510

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point ≥142 °C
Assay ≥96.0 %
Loss on drying ≤0.5 %
Additional info additive: 0.5 to 2.5% oil
Particle size ≤0.1 % Residue on 150 micron sieve
Ash ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR