Didecylsulfid ≥97%

Lieferant: Thermo Scientific

B22560.09 B22560.18
B22560.09EA 27.7 EUR
B22560.09 B22560.18
Didecylsulfid ≥97%
Didecylsulfid

Purity: 97% CAS No.: 693-83-4 Molecular Formula: [CH3(CH2)9]2S Formula Weight: 314.63 Melting Point: 25-28° Boiling Point: 217-218°/8mm MDL No.: MFCD00026542

Formel: C₂₀H₄₂S
Molecular Weight: 314,62 g/mol
Siedepunkt: 217…218 °C (8 mmHg)
Schmelzpunkt: 25…28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00026542
CAS-Nummer: 693-83-4
EINECS: 211-761-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR