Dihexylsulfid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Di-n-hexylsulphide
B22325.14
B22325.22
B22325.14EA
45.2
EUR
B22325.14
B22325.22
Dihexylsulfid ≥97%
Dihexylsulfid
Purity: 97%
CAS No.: 6294-31-1
Molecular Formula: [CH3(CH2)5]2S
Formula Weight: 202.40
Flash Point: 116°(241°F)
Refractive Index: 1.4587
Odour: Stench
Boiling Point: 260°
Beilstein Reference: 1738333
UN No.: 3334
MDL No.: MFCD00009528
Formel:
C₁₂H₂₆S Molecular Weight: 202,4 g/mol Siedepunkt: 260 °C Schmelzpunkt: –28,3 °C Dichte: 0,849 Flammpunkt: 116 °C (241 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009528 CAS-Nummer: 6294-31-1 EINECS: 228-556-1 UN: 3334 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...