Dimethyldisulfid ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Dimethyl disulphide, Methyl disulphide

L09679.30 L09679.0B
L09679.30EA 31.5 EUR
L09679.30 L09679.0B
Dimethyldisulfid ≥99%
Dimethyldisulfid

Purity: 99% CAS No.: 624-92-0 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 10°(50°F) Refractive Index: 1.5253 Odour: Stench Melting Point: -85° Boiling Point: 108-109° Beilstein Reference: 1730824 UN No.: 2381 MDL No.: MFCD00008561

Formel: C₂H₆S₂
Molecular Weight: 94,2 g/mol
Siedepunkt: 108…109 °C
Schmelzpunkt: –85 °C
Dichte: 1,046
Flammpunkt: 10 °C (50 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008561
CAS-Nummer: 624-92-0
EINECS: 210-871-0
UN: 2381
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR