Dimethylsulfid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
DMS
22949.AK
22949.K2
22949.AKEA
40.5
EUR
22949.AK
22949.K2
Dimethylsulfid ≥99%
Dimethylsulfid
Purity: 99+%
CAS No.: 75-18-3
Molecular Formula: (CH3)2S
Formula Weight: 62.14
Flash Point: -48°(-54°F)
Physical Form: Liquid
Density: 0.845
Refractive Index: 1.4351
Odour: Unpleasant
Melting Point: -83°
Boiling Point: 38°
Merck Reference: 14,6123
UN No.: 1164
MDL No.: MFCD00008562
Formel:
C₂H₆S Molecular Weight: 62,14 g/mol Siedepunkt: 38 °C Schmelzpunkt: –83 °C Dichte: 0,845 Flammpunkt: –48 °C (–54 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008562 CAS-Nummer: 75-18-3 EINECS: 200-846-2 UN: 1164 ADR: 3,II Merck Index: 14,6123 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...