Dioctylsulfid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dioctylsulphane

A17955.14 A17955.22 A17955.36
A17955.14EA 45.8 EUR
A17955.14 A17955.22 A17955.36
Dioctylsulfid ≥97%
Dioctylsulfid

Purity: 97% CAS No.: 2690-08-6 Molecular Formula: [CH3(CH2)7]2S Formula Weight: 258.51 Flash Point: 160°(320°F) Refractive Index: 1.4624 Odour: Stench Melting Point: 0-1° Boiling Point: 179-180°/10mm Beilstein Reference: 1701624 MDL No.: MFCD00009566

Formel: C₁₆H₃₄S
Molecular Weight: 258,51 g/mol
Siedepunkt: 179…180 °C (10 mmHg)
Schmelzpunkt: 0…1 °C
Dichte: 0,844
Flammpunkt: 160 °C (320 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009566
CAS-Nummer: 2690-08-6
EINECS: 220-259-5
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR