Dipropyldisulfid ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 629-19-6
Molecular Formula: CH3CH2CH2S-SCH2CH2CH3
Formula Weight: 150.31
Flash Point: 64°(147°F)
Refractive Index: 1.4967
Odour: Unpleasant
Melting Point: -86°
Boiling Point: 193-194°
Beilstein Reference: 969200
UN No.: 3082
MDL No.: MFCD00009378
Formel:
C₆H₁₄S₂ Siedepunkt: 193…194 °C Schmelzpunkt: –86 °C Dichte: 0,960 Flammpunkt: 64 °C (147 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009378 CAS-Nummer: 629-19-6 EINECS: 211-079-8 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...