Iodmonochlorid ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Iodochloride
44348.14
44348.22
44348.36
44348.14EA
52.4
EUR
44348.14
44348.22
44348.36
Iodmonochlorid ACS
Iodmonochlorid
CAS No.: 7790-99-0
Molecular Formula: ClI
Formula Weight: 162.36
Physical Form: Crystalline/Liquid
Density: 3.182
Sensitivity: Moisture and Light Sensitive
Appearance: Dark red/black
Odour: Pungent
Melting Point: 25-27°
Boiling Point: 96-98°
Beilstein Reference: 3902972
Merck Reference: 14,5017
UN No.: 1792
MDL No.: MFCD00011354
Formel:
ClI Molecular Weight: 162,36 g/mol Siedepunkt: 96…98 °C Schmelzpunkt: 25…27 °C Dichte: 3,182 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011354 CAS-Nummer: 7790-99-0 EINECS: 232-236-7 UN: 1792 ADR: 8,II Merck Index: 14,5017 |