1,1'-Carbonyldiimidazol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,1'-Carbonylbis(imidazol), 1,1'-Carbonylbis-1H-imidazol, N,N'-Carbonyldiimidazol, CDI
A14688.30
A14688.09
A14688.18
A14688.30EA
396
EUR
A14688.30
A14688.09
A14688.18
1,1'-Carbonyldiimidazol ≥97%
1,1'-Carbonyldiimidazol
Purity: 97%
CAS No.: 530-62-1
Molecular Formula: C7H6N4O
Formula Weight: 162.15
Physical Form: Cryst powder
Sensitivity: Moisture Sensitive
Appearance: White to cream
pH: 7.7
Melting Point: 116-120°
Beilstein Reference: 6826
Merck Reference: 14,1819
UN No.: 3263
MDL No.: MFCD00005286
Peptide coupling reagent
Formel:
C₇H₆N₄O Molecular Weight: 162,15 g/mol Schmelzpunkt: 116…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005286 CAS-Nummer: 530-62-1 EINECS: 208-488-9 UN: 3263 ADR: 8,II Merck Index: 14,1819 |