1,1'-Carbonyldiimidazol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
1,1'-Carbonylbis(imidazol), 1,1'-Carbonylbis-1H-imidazol, N,N'-Carbonyldiimidazol, CDI
003800-1KG
003800-250G
003800-50G
FLUO003800-1KGEA
0
EUR
FLUO003800-1KG
FLUO003800-250G
FLUO003800-50G
1,1'-Carbonyldiimidazol
1,1'-Carbonyldiimidazol
Formel:
C₇H₆N₄O Molecular Weight: 162,15 g/mol Schmelzpunkt: 111…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005286 CAS-Nummer: 530-62-1 EINECS: 208-488-9 UN: 3263 ADR: 8,II Merck Index: 14,01819 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...