4-Phenylimidazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Phenyl-1H-imidazole
L02237.04
L02237.09
L02237.04EA
47.1
EUR
L02237.04
L02237.09
4-Phenylimidazol ≥98%
4-Phenylimidazol
Purity: 98+%
CAS No.: 670-95-1
Molecular Formula: C9H8N2
Formula Weight: 144.18
Melting Point: 143-145°
Beilstein Reference: 2969
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Siedepunkt: 182…184 °C (1 mmHg) Schmelzpunkt: 143…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005197 CAS-Nummer: 670-95-1 EINECS: 211-580-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...