4-Phenylimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Phenyl-1H-imidazole
144754-10G
144754-5G
SIAL144754-10GEA
98.4
EUR
SIAL144754-10G
SIAL144754-5G
4-Phenylimidazol, Sigma-Aldrich®
4-Phenylimidazol
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Siedepunkt: 182…184 °C (1 mmHg) Schmelzpunkt: 144…146 °C |
MDL:
MFCD00005197 CAS-Nummer: 670-95-1 EINECS: 211-580-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...