2-Brom-1-indanon
Lieferant: CHEMPUR
|
Achtung
|
CD00348.250
CD00348.1
CHMPCD00348.250EA
0
EUR
CHMPCD00348.250
MAYBCD00348.1
2-Brom-1-indanon
2-Brom-1-indanon
Formel:
C₉H₇BrO Molecular Weight: 211,06 g/mol Siedepunkt: 110…112 °C (0,4 mmHg) Schmelzpunkt: 38 °C |
MDL:
MFCD00041030 CAS-Nummer: 1775-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...