5-Brom-1-indanon

Lieferant: CHEMPUR

Synonyme: 5-Bromo-2,3-dihydro-1H-inden-1-one

BTB14082.250 BTB14082.1
CHMPBTB14082.250EA 0 EUR
CHMPBTB14082.250 MAYBBTB14082.1
5-Brom-1-indanon
5-Brom-1-indanon
Formel: C₉H₇BrO
Molecular Weight: 211,06 g/mol
Siedepunkt: 280 °C (1013 hPa)
Schmelzpunkt: 126…127 °C
MDL: MFCD00082718
CAS-Nummer: 34598-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR