4-Bromindol ≥98%
Lieferant: Thermo Scientific
Synonyme:
4-Bromo-1H-indole
L19417.03
L19417.06
L19417.03EA
64.4
EUR
L19417.03
L19417.06
4-Bromindol ≥98%
4-Bromindol
Purity: 98%
CAS No.: 52488-36-5
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₈H₆BrN Molecular Weight: 196,05 g/mol Siedepunkt: 88…90 °C (0,1 mmHg) Schmelzpunkt: 17 °C Dichte: 1,563 Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00671502 CAS-Nummer: 52488-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...