4-Bromindol

Lieferant: CHEMPUR

Synonyme: 4-Bromo-1H-indole

FL13448.POR 013448-1G 013448-5G
CHMPFL13448.POREA 0 EUR
CHMPFL13448.POR FLUO013448-1G FLUO013448-5G
4-Bromindol
4-Bromindol
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
MDL: MFCD00671502
CAS-Nummer: 52488-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR