6-Bromindol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Bromo-1H-indole
L17703.03
L17703.MD
L17703.03EA
148
EUR
L17703.03
L17703.MD
6-Bromindol ≥98%
6-Bromindol
Purity: 98%
CAS No.: 52415-29-9
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Melting Point: 91-93°
Boiling Point: 70-75°/0.01mm
MDL No.: MFCD00238550
Formel:
C₈H₆BrN Molecular Weight: 196,05 g/mol Siedepunkt: 70…75 °C (0,01 mmHg) Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238550 CAS-Nummer: 52415-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...