6-Bromindol

Lieferant: Thermo Scientific

Synonyme: 6-Bromo-1H-indole

RH00739.250 RH00739.1
MAYBRH00739.250EA 0 EUR
MAYBRH00739.250 MAYBRH00739.1
6-Bromindol
6-Bromindol
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00238550
CAS-Nummer: 52415-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR