5-Chlorindol 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Chloro-1H-indole
109310010
109310050
ACRO109310010EA
52.9
EUR
ACRO109310010
ACRO109310050
5-Chlorindol 99%
5-Chlorindol
Formel:
C₈H₆ClN Molecular Weight: 151,6 g/mol Schmelzpunkt: 69…72 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005672 CAS-Nummer: 17422-32-1 EINECS: 241-448-9 UN: 2811 |
Spezifikation Testergebnisse
Appearance | White to grey-green Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 69 to 72 °C |
GC | ≤22 % (alpha-lactose) |
Sulfated ash | ≤0.1 % |
Appearance of solution | (per Ph. Eur.) Passes test |
Microbial contamination | (per Ph. Eur.) Passes test |
Acidity/alkalinity | ≤0.4 ml/6g (ml 0.1N NaOH/6g) |
Specific optical rotation | +54.4° to +55.9° (20 °C, 589 nm) (c=10, H2O/NH3) (on anhydrous basis) |
Absorbance | (per Ph. Eur.) Passes test |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...