Indol-3-carbonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Cyan-1H-indol, 3-Indolcarbonitril, 1H-Indol-3-carbonitril, 3-Cyanindol
A16041.03
A16041.06
A16041.03EA
46.6
EUR
A16041.03
A16041.06
Indol-3-carbonitril ≥98%
Indol-3-carbonitril
Purity: 98%
CAS No.: 5457-28-3
Molecular Formula: C9H6N2
Formula Weight: 142.16
Odour: Odourless
Melting Point: 177-182°
Beilstein Reference: 120888
MDL No.: MFCD00022717
Formel:
C₉H₆N₂ Siedepunkt: 350 °C (1013 hPa) Schmelzpunkt: 177…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022717 CAS-Nummer: 5457-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...