Indol-5-carbonitril ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1H-Indol-5-carbonitril

B21810.03 B21810.06
B21810.03EA 40.8 EUR
B21810.03 B21810.06
Indol-5-carbonitril ≥98%
Indol-5-carbonitril

Purity: 98+% CAS No.: 15861-24-2 Molecular Formula: C9H6N2 Formula Weight: 142.16 Odour: Odourless Melting Point: 104-108° Beilstein Reference: 116738 MDL No.: MFCD00005669

Formel: C₉H₆N₂
Molecular Weight: 142,16 g/mol
Schmelzpunkt: 104…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005669
CAS-Nummer: 15861-24-2
EINECS: 239-986-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR