5-Indolcarbonsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1H-Indole-5-carboxylic acid
122210010
122210050
ACRO122210010EA
38.7
EUR
ACRO122210010
ACRO122210050
5-Indolcarbonsäure 98%
5-Indolcarbonsäure
Formel:
C₉H₇NO₂ Schmelzpunkt: 207…212 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005678 CAS-Nummer: 1670-81-1 EINECS: 216-799-6 |
Spezifikation Testergebnisse
Appearance | Yellow to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 207 to 212 °C |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...