4-Fluorindol

Lieferant: CHEMPUR
Achtung

48-0009.1 48-0009.5
CHMP48-0009.1EA 0 EUR
CHMP48-0009.1 CHMP48-0009.5
4-Fluorindol
4-Fluorindol
Formel: C₈H₆FN
MDL: MFCD00055992
CAS-Nummer: 387-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR