6-Fluorindol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Fluoro-1H-indole

B24633.03 B24633.06 B24633.14
B24633.03EA 59.9 EUR
B24633.03 B24633.06 B24633.14
6-Fluorindol ≥98%
6-Fluorindol

Purity: 98% CAS No.: 399-51-9 Molecular Formula: C8H6FN Formula Weight: 135.14 Melting Point: 74-77° Beilstein Reference: 112192

Formel: C₈H₆FN
Molecular Weight: 135.14 g/mol
Schmelzpunkt: 74…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00056933
CAS-Nummer: 399-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR