7-Fluorindol

Lieferant: FLUOROCHEM
Achtung

CFF1111. 005411-1G 005411-5G
CHMPCFF1111.EA 0 EUR
CHMPCFF1111. FLUO005411-1G FLUO005411-5G
7-Fluorindol
7-Fluorindol
Formel: C₈H₆FN
Molecular Weight: 135.14 g/mol
MDL: MFCD01074502
CAS-Nummer: 387-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR