5-Iodindol

Lieferant: CHEMPUR
Achtung

Synonyme: 5-Jodindol, 5-Jod-1H-indol

RH00726.250 RH00726.1
CHMPRH00726.250EA 0 EUR
CHMPRH00726.250 MAYBRH00726.1
5-Iodindol
5-Iodindol
Formel: C₈H₆IN
Molecular Weight: 243,05 g/mol
MDL: MFCD00220065
CAS-Nummer: 16066-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR