5-Iodindol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Jodindol, 5-Jod-1H-indol

M-Clarity™ Qualitätsstufe = MQ100

563838-25G 563838-5G
SIAL563838-25GEA 438 EUR
SIAL563838-25G SIAL563838-5G
5-Iodindol, Sigma-Aldrich®
5-Iodindol
Formel: C₈H₆IN
Molecular Weight: 243,05 g/mol
MDL: MFCD00220065
CAS-Nummer: 16066-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR