4-Nitroindol

Lieferant: Thermo Scientific

Synonyme: 4-Nitro-1H-indole

RH01152.250 RH01152.1
MAYBRH01152.250EA 0 EUR
MAYBRH01152.250 MAYBRH01152.1
4-Nitroindol
4-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 206…209 °C
MDL: MFCD00010056
CAS-Nummer: 4769-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR