5-Nitroindol

Lieferant: FLUOROCHEM

Synonyme: 5-Nitro-1H-indole

010352-5G 010352-25G
FLUO010352-5GEA 34.1 EUR
FLUO010352-5G FLUO010352-25G
5-Nitroindol
5-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 140 °C
MDL: MFCD00005673
CAS-Nummer: 6146-52-7
EINECS: 228-153-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR