7-Nitroindol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

638854-1G 638854-5G
SIAL638854-1GEA 49.2 EUR
SIAL638854-1G SIAL638854-5G
7-Nitroindol, Sigma-Aldrich®
7-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD00005683
CAS-Nummer: 6960-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR